Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIK3C3 (phospho S164), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PIK3C3.
Supplier: Abnova Corporation PAB8083
Specifications
PIK3C3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PIK3C3. | |
In PBS (0.09% sodium azide) | |
MGC61518/Vps34 | |
Rabbit | |
Protein A purification | |
RUO | |
5289 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Dot Blot, ELISA | |
Unconjugated | |
ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
PIK3C3 | |
PIK3C3 | |
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S164 of human PIK3C3. | |
400 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction