Learn More
PIP5K3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PIP5K3.
Supplier: Abnova Corporation PAB2729
Description
PIP5K3 belongs to a large family of lipid kinases that alter the phosphorylation status of intracellular phosphatidylinositol. Signaling by phosphorylated species of phosphatidylinositol regulates diverse cellular processes, including membrane trafficking and cytoskeletal reorganization (Shisheva et al., 1999 [PubMed 9858586]).[supplied by OMIM
Specifications
PIP5K3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PIP5K3. | |
In PBS (0.09% sodium azide) | |
CFD/FAB1/KIAA0981/MGC40423/PIKFYVE/PIP5K | |
Rabbit | |
Protein G purification | |
RUO | |
200576 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
PIP5K3 | |
PIP5K3 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human PIP5K3. | |
400 μL | |
Primary | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only