Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIWIL4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant PIWIL4.
Supplier: Abnova Corporation PAB18860
Description
PIWIL4 belongs to the Argonaute family of proteins, which function in development and maintenance of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).[supplied by OMIM
Specifications
PIWIL4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant PIWIL4. | |
In buffer containing 0.02% sodium azide, 2% BSA | |
DKFZp686P01248/FLJ36156/HIWI2/MIWI2 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:5000-1:20000) Western Blot (1:200-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
PIWIL4 | |
PIWIL4 | |
Recombinant protein corresponding to human PIWIL4. | |
RUO | |
143689 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction