Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rabbit anti-PKC delta, Clone: K15-K, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP1301250.05ML
Description
PKC delta Monoclonal specifically detects PKC delta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
PKC delta | |
Monoclonal | |
0.5mg/mL | |
Western Blot 1:1000, ELISA 1:5000-1:10000, Immunoprecipitation 1:10-1:500 | |
Q05655 | |
PRKCD | |
Peptide derived from the human PKC-delta sequence | |
0.05 mL | |
Primary | |
This antibody is specific for PKC-delta. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunoprecipitation | |
K15-K | |
Unconjugated | |
20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide | |
EC 2.7.11, EC 2.7.11.13, MAY1, MGC49908, nPKC-delta, PKCD, protein kinase C delta type, protein kinase C delta VIII, protein kinase C, delta | |
Rabbit | |
Affinity Purified | |
RUO | |
5580 | |
Human, Mouse, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction