Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA2G4E, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PLA2G4E.
Manufacturer: Abnova Corporation PAB17689
Specifications
PLA2G4E | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PLA2G4E. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
FLJ45651/MGC126633/MGC126661 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total PLA2G4E protein. | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
PLA2G4E | |
PLA2G4E | |
A synthetic peptide corresponding to internal of human PLA2G4E. | |
RUO | |
123745 | |
Human |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only