Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PNN.
Supplier: Abnova Corporation PAB28421
Description
Sequence: EQKVELAQLQEEWNEHNAKIIKYIRTKTKPHLFYIPGRMCPATQKLIEESQRKMNALFEGRRIEFAEQINKMEARPRRQSMKEKEHQVVRNEEQKAEQEEGKVAQREEELEETGNQHNDVEIEEAGEEEEKEIAIVHSDAEKESpecifications
PNN | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant PNN. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
DRS/SDK3/memA/pinin | |
Rabbit | |
Antigen affinity purification | |
RUO | |
5411 | |
Store at 4 °C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:250-1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
PNN | |
PNN | |
Recombinant protein corresponding to human PNN. | |
100 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only