Learn More
PNOC Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic protein of PNOC.
Manufacturer:  Abnova Corporation PAB19565
Catalog No. 89-121-093
Add to cart
Specifications
Specifications
PNOC
Unconjugated
ELISA (1-2 ug/mL) Western Blot (2-10 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user.
PNOC
PPNOC
Rabbit
Protein G purification
RUO
Primary
5368
ELISA, Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic protein of PNOC.
Lyophilized from PBS.
Lyophilized
PNOC
Synthetic protein corresponding to human PNOC.
100 ug
Store at -20°C on dry atmosphere. After reconstitution with deionized water
Polyclonal
Human, Murine, Rabbit, Rat