Learn More
POLR2J, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of POLR2J.
Supplier: Abnova Corporation PAB17697
Description
This gene encodes a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene exists as a heterodimer with another polymerase subunit; together they form a core subassembly unit of the polymerase. Two similar genes are located nearby on chromosome 7q22.1 and a pseudogene is found on chromosome 7p13. [provided by RefSeq
Specifications
POLR2J | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of POLR2J. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
MGC71910/POLR2J1/RPB11/RPB11A/RPB11m/hRPB14 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total POLR2J protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
POLR2J | |
POLR2J | |
A synthetic peptide corresponding to internal of human POLR2J. | |
RUO | |
5439 | |
Human, Mouse | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only