Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR gamma/NR1C3 (p Ser273) Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Canine, Primate, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244159

Catalog No. NBP244159


Add to cart

Description

Description

Specifically detects PPAR gamma/NR1C3 in Human samples, and it is validated for ELISA, Western Blotting

PPAR gamma/NR1C3 (p Ser273) Polyclonal antibody specifically detects PPAR gamma/NR1C3 (p Ser273) in Human, Mouse, Rat, Porcine, Bovine, Canine, Primate samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PPAR gamma/NR1C3 (p Ser273)
0.85 mg/mL
Western Blot 1ug/ml, ELISA 1: 10000
Affinity Purified
PPARG
Anti-PPAR gamma [p Ser273] affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the internal region of human PPARG protein.
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat, Porcine, Bovine, Canine, Primate
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.05% Sodium Azide
CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Rabbit
IgG
0.05 mg
Cancer, mTOR Pathway, Phospho Specific
Primary
5468
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only