Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R2A Rabbit anti-Bovine, Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of PPP2R2A.

Manufacturer:  Abnova Corporation PAB13657

Catalog No. 89-137-649


Add to cart

Description

Description

Sequence: FSQVKGAVDDDVAE
Specifications

Specifications

PPP2R2A
Unconjugated
Western Blot (5-10 ug/mL) Immunocytochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide (conjugated with KLH) corresponding to rabbit PPP2R2A.
RUO
Primary
This is a rabbit polyclonal antibody is specific to serine/threonine proten phosphatase 2 A/Balpha (PP 2 A/Balpha ) corresponding to a specific peptide immunogen to enhance reactivity and minimize cross reactivity. This antibody does not cross-react with other protein phosphatase isoforms.
Immunocytochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of PPP2R2A.
In PBS (0.08% sodium azide)
Rabbit
100 ug
Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Bovine, Human, Murine, Rabbit, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only