Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRKCB (phospho S660), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKCB.

Manufacturer:  Abnova Corporation PAB17906

Catalog No. 89-119-546


Add to cart

Description

Description

Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase has been reported to be involved in many different cellular functions, such as B cell activation, apoptosis induction, endothelial cell proliferation, and intestinal sugar absorption. Studies in mice also suggest that this kinase may also regulate neuronal functions and correlate fear-induced conflict behavior after stress. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq

Specifications

Specifications

PRKCB
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
PRKCB
MGC41878|PKC-beta|PKCB|PRKCB1|PRKCB2
Rabbit
100 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody recognizes several Phosphorylated PKC (Ser660) isoforms of human, mouse, and rat origins.
Western Blot
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKCB.
In PBS (50% glycerol, 1% BSA, 0.02% sodium azide)
Liquid
PRKCB
Synthetic phosphopeptide corresponding to residues surrounding S660 of human PRKCB.
RUO
Primary
5579
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only