Learn More
PRKD1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRKD1.
Supplier:  Abnova Corporation PAB4541
Catalog No. 89-131-455
Add to Cart
Description
Description
Members of the protein kinase C (PKC) family function in many extracellular receptor-mediated signal transduction pathways. See PRKCA (MIM 176960) for further background information. The PRKCM gene encodes a cytosolic serine-threonine kinase that binds to the trans-Golgi network and regulates the fission of transport carriers specifically destined to the cell surface.[supplied by OMIM
Specifications
Specifications
PRKD1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of PRKD1.
In PBS (0.09% sodium azide)
PKC-MU/PKCM/PKD/PRKCM
Rabbit
Protein G purification
RUO
5587
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
PRKD1
PRKD1
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PRKD1.
400 μL
Primary
Human, Mouse
Liquid
Product Suggestions
Product Suggestions