Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proBDNF Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244063

Catalog No. NBP244063


Add to cart

Description

Description

Specifically detects proBDNF in C. elegans, Human samples, and it is validated for ELISA, Western Blotting

proBDNF Polyclonal antibody specifically detects proBDNF in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

proBDNF
LYOPH
Western Blot 1:1000, ELISA 1:10000
Purified
PZP
This IgG fraction antibody was prepared from rabbit antiserum after repeated immunizations with recombinant truncated human proBDNF protein produced in E.coli.
IgG purified
RUO
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 100 ul deionized water.
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with No Preservative
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Rabbit
IgG
0.1 mg
Cytokine Research, Immunology, Neuroscience
Primary
627
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only