Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Prokineticin 2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243853

Catalog No. NBP243853


Add to cart

Description

Description

Specifically detects Prokineticin 2 in Bovine, Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immunofluorescence, Western Blotting

Prokineticin 2 Polyclonal antibody specifically detects Prokineticin 2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Prokineticin 2
LYOPH
Western Blot 1:200-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
PROK2
Synthetic peptide derived from Cter region of human PK2
Protein A purified
RUO
Lyophilized antibody can be kept at 4C for up to 3 months and should be kept at -20C for long-term storage. To avoid freeze-thaw cycles, reconstituted antibody should be aliquoted before freezing for short-term storage (-20C) or for long-term storage (-80C). For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer.
Polyclonal
Reconstitute with 0.1ml of distilled water.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
0.1M Tris 0.1M Glycine and 2% Sucrose with No Preservative
BV8Protein Bv8 homolog, KAL4, MIT1, PK2prokineticin-2, prokineticin 2
Rabbit
IgG
0.1 mg
Angiogenesis
Primary
60675
Human, Mouse, Rat, Bovine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only