Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROKR2 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of PROKR2.

Manufacturer:  Abnova Corporation PAB25994

Catalog No. 89-127-630


Add to cart

Description

Description

Prokineticins are secreted proteins that can promote angiogenesis and induce strong gastrointestinal smooth muscle contraction. The protein encoded by this gene is an integral membrane protein and G protein-coupled receptor for prokineticins. The encoded protein is similar in sequence to GPR73, another G protein-coupled receptor for prokineticins. [provided by RefSeq

Specifications

Specifications

PROKR2
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15-30 ug/mL) The optimal working dilution should be determined by the end user.
PROKR2
GPR73L1|GPR73b|GPRg2|KAL3|PKR2|dJ680N4.3
Rabbit
Immunoaffinity chromatography
RUO
Primary
128674
Human, Monkey
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of PROKR2.
In PBS (0.09% sodium azide)
Liquid
PROKR2
A synthetic peptide corresponding to 16 amino acids at C-terminus of human PROKR2.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
BLAST analysis of the peptide immunogen showed no homology with other human proteins, except PROKR1 (44%).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only