Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Prolactin, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP1363880.1ML

Catalog No. NBP13638801

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Prolactin
Unconjugated
10mM PBS (pH 7.4) with 0.1% Sodium Azide
prolactin
Rabbit
IgG
0.1 ml
Cancer
Primary
5617
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Affinity Purified
PRL
Purified human prolactin
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
This product is specific for Prolactin
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit