Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCH1 Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of PTCH1.

Manufacturer:  Abnova Corporation PAB12543

Catalog No. 89-115-913


Add to cart

Description

Description

Specifications

Specifications

PTCH1
Unconjugated
ELISA (1:500-1:1000) Immunocytochemistry (1:500-1:1000) Immunohistochemistry (Frozen sections) (1:500-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:1000) Western Blot (1:500-1:1000) The optimal workin
Liquid
PTCH1
A synthetic peptide corresponding to amino acids 269-279 of human and mouse PTCH1/Ptch1.
RUO
Primary
5727|19206
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of PTCH1.
In PBS (0.02% sodium azide)
BCNS|FLJ26746|FLJ42602|HPE7|NBCCS|PTC|PTC1|PTCH|PTCH11
Rabbit
100 uL
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Chicken, Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only