Learn More
PTGER2 Rabbit anti-Bovine, Human, Mouse, Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTGER2.
Manufacturer:  Abnova Corporation PAB16359
Catalog No. 89-118-229
Add to cart
Description
Description
Specifications
Specifications
PTGER2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of PTGER2.
In PBS (0.09% sodium azide)
EP2
Rabbit
Immunoaffinity purification
RUO
5732
Bovine, Human, Mouse, Porcine
Liquid
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (40 ug/mL) The optimal working dilution should be determined by the end user.
PTGER2
PTGER2
A synthetic peptide (conjugated with KLH) corresponding to human PTGER2.
50 μg
Primary
3rd extracellular domain of human.
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.