Learn More
PTK2 Rabbit anti-Bovine, Chicken, Dog, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTK2.
Manufacturer: Abnova Corporation PAB16323
Description
This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. At least four transcript variants encoding four different isoforms have been found for this gene, but the full-length natures of only two of them have been determined. [provided by RefSeq
Specifications
PTK2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PTK2. | |
In PBS (0.09% sodium azide) | |
FADK/FAK/FAK1/pp125FAK | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
5747 | |
Bovine, Canine, Chicken, Equine, Hamster, Human, Monkey, Murine, Rabbit, Rat |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2 ug/mL) The optimal working dilution should be determined by the end user. | |
PTK2 | |
PTK2 | |
A synthetic peptide (conjugated with KLH) corresponding to human PTK2. | |
50 μg | |
Primary | |
Internal domain of human. | |
Liquid |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only