Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTN Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PTN.
Supplier: Abnova Corporation PAB19496
Specifications
PTN | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PTN. | |
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
HARP/HBGF8/HBNF/NEGF1 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
5764 | |
Human, Rat | |
Lyophilized |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1 ug/mL) The optimal working dilution should be determined by the end user. | |
PTN | |
PTN | |
A synthetic peptide corresponding to amino acids at C-terminus of human PTN. | |
100 μg | |
Primary | |
It identical to the rat and mouse sequences. | |
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction