Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB5A, Rabbit, HRP, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.
Supplier: Abnova Corporation PAB29013
Specifications
RAB5A | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of RAB5A. | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
RAB5 | |
Rabbit | |
Affinity purification | |
RUO | |
5868 | |
Bovine, Human, Monkey, Mouse, Rat | |
Liquid |
Immunofluorescence, Western Blot | |
HRP | |
Immunofluorescence Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
RAB5A | |
RAB5A | |
A synthetic peptide conjugated with KLH corresponding to human RAB5A. | |
100 μg | |
Primary | |
The antibody recognizes 26 kDa Rab5 of the above species origins. | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only