Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-RABGAP1, (1 to 50), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244284

Catalog No. NBP244284


Add to cart

Description

Description

Specifically detects RABGAP1 in Human samples, and it is validated for Western Blotting

RABGAP1 Polyclonal antibody specifically detects RABGAP1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RABGAP1
1.0 mg/mL
Western Blot 1:1000 - 1:5000
Affinity Purified
RABGAP1
The epitope recognized by this antibody maps to a region between residue 1 to 50 of human Rab GTPase-activating protein 1 using the numbering given in entry Q9Y3P9.3 (GeneID 23637).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
DKFZp586D2123, GAP and centrosome-associated protein, GAPCENA, RAB GTPase activating protein 1, rab GTPase-activating protein 1, rab6 GTPase activating protein (GAP and centrosome-associated), Rab6 GTPase activating protein, GAPCenA, Rab6 GTPase-activating protein GAPCenA, RP11-123N4.2, TBC1 domain family, member 11, TBC1D11
Rabbit
IgG
100 ul
Cell Cycle and Replication
Primary
23637
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only