Learn More
RANBP10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of RANBP10.
Manufacturer:  Abnova Corporation PAB19220
Catalog No. 89-120-750
Add to cart
Description
Description
Specifications
Specifications
RANBP10
1 mg/mL
Rabbit polyclonal antibody rasied against synthetic peptide of RANBP10.
In PBS (0.02% sodium azide)
Liquid
RANBP10
A synthetic peptide corresponding to 15 amino acids near internal region of human RANBP10.
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Immunohistochemistry, Western Blot
Unconjugated
Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
RANBP10
FLJ31165|KIAA1464
Rabbit
Peptide affinity purification
RUO
Primary
57610