Learn More
IgG, IgM, IgA (H+L) Rabbit anti-Rat, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Manufacturer:  Invitrogen SA136147
Catalog No. 01-671-522
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Thermo Scientific Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Rat IgG, IgM, IgA (H+L)
Polyclonal
Unconjugated
Liquid
Immunoglobulins.
Purified
RUO
Secondary Antibody
Polyclonal
Dot Blot, ELISA
1 mg/mL
0.01M sodium phosphate with 0.1M NaCl, 50% glycerol and no preservative; pH 7.5
Rabbit
IgG
1 mL
4° C
Secondary
Rat