Learn More
RCAN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RCAN1.
Supplier:  Abnova Corporation PAB4386
Catalog No. 89-131-396
Add to Cart
Description
Description
The protein encoded by this gene interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses. Chronic overexpression of this gene may lead to neurofibrillary tangles such as those associated with Alzheimer disease. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
RCAN1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of RCAN1.
In PBS (0.09% sodium azide)
ADAPT78/CSP1/DSC1/DSCR1/MCIP1/RCN1
Rabbit
Protein G purification
RUO
1827
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user.
RCAN1
RCAN1
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human RCAN1.
400 μL
Primary
Human
Liquid
Recommendations
Recommendations