Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REL Rabbit anti-Flies, Fruit fly, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Rel.

Manufacturer:  Abnova Corporation PAB14193

Catalog No. 89-116-978

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

REL
Unconjugated
ELISA (1:1000-1:5000) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Rel
CG11992|ird|ird4|l(3)neo36|unnamed
Rabbit
200 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Polyclonal
This antibody detects specifically target peptide derived from Drosophila Relish by ELISA and Dot Blotting. Also detects cleaved and uncleaved Relish in PFA-fixed Drosphila tissue (see image). Cross reactivity with human, mouse and rat have not been tested.
ELISA, Immunofluorescence, Immunohistochemistry
Rabbit polyclonal antibody raised against synthetic peptide of Rel.
1X PBS (pH 7.4)
Lyophilized
Rel
A synthetic peptide (conjugated with KLH) corresponding to Drosophila Rel.
RUO
Primary
41087
Drosophila, Fly
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only