Learn More
REL Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of REL.
Supplier: Abnova Corporation PAB4433
Description
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA (MIM 164014), RELB (604758), NFKB1 (MIM 164011), and NFKB2 (MIM 164012). These proteins are related through a highly conserved N-terminal region termed the 'Rel domain,' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (MIM 164008) (Belguise and Sonenshein, 2007 [PubMed 18037997]).[supplied by OMIM
Specifications
REL | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of REL. | |
In PBS (0.09% sodium azide) | |
C-Rel | |
Rabbit | |
Protein G purification | |
RUO | |
5966 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
REL | |
REL | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human REL. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only