Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Renilla Luciferase, (aa 1-311), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242914

Catalog No. NBP242914


Add to cart

Description

Description

Specifically detects Renilla Luciferase in Human, Invertebrate samples, and it is validated for Western Blotting

Renilla Luciferase Polyclonal antibody specifically detects Renilla Luciferase in Invertebrate samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Renilla Luciferase
Unconjugated
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
Invertebrate
Western Blot, Immunohistochemistry, Immunohistochemistry Whole-Mount
Western Blot 1:500-1:3000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry Whole-Mount
Affinity Purified
The exact sequence is proprietary.
36 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only