Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RETNLB Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RETNLB.
Manufacturer: Abnova Corporation PAB18038
Specifications
RETNLB | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of RETNLB. | |
In PBS, pH 7.5, 0.1% BSA. | |
FIZZ1/FIZZ2/HXCP2/RELM-beta/RELMb/RELMbeta/XCP2 | |
Rabbit | |
200 μg | |
Primary | |
The rabbit anti-human RETNLB antibody cross reactivity with mouse and rat are expected from sequence similarity. | |
Lyophilized |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000-1:5000) Western Blot (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
RETNLB | |
RETNLB | |
A synthetic peptide corresponding to human RETNLB. | |
RUO | |
84666 | |
Human, Mouse, Rat |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only