Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-RIC8A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236536

Catalog No. NBP236536


Add to cart

Description

Description

RIC8A Polyclonal antibody specifically detects RIC8A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

RIC8A
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg
EAW61245.1
RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
RIC8A
The immunogen recognized by the antibody maps to a region between residue 418 to 468 of human Resistance to Inhibitors of Cholinesterase 8 Homolog A using the numbering given in entry EAW61245.1.
Immunogen affinity purified
RUO
Primary
60626
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit