Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIT1, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of RIT1.

Supplier:  Abnova Corporation PAB27087

Catalog No. 89-128-738


Add to Cart

Description

Description

RIT belongs to the RAS (HRAS; MIM 190020) subfamily of small GTPases (Hynds et al., 2003 [PubMed 12668729]).[supplied by OMIM

Specifications

Specifications

RIT1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of RIT1.
In PBS, pH 7.2 (0.09% sodium azide)
MGC125864/MGC125865/RIBB/RIT/ROC1
Rabbit
Antigen affinity purification
RUO
6016
Human, Mouse
Liquid
Western Blot
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
RIT1
RIT1
A synthetic peptide corresponding to human RIT1.
100 μL
Primary
RIT1 polyclonal antibody detects endogenous levels of RIT1 protein.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only