Learn More
RMI2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RMI2.
Supplier: Abnova Corporation PAB23731
Description
RMI2 is a component of the BLM (RECQL3; MIM 604610) complex, which plays a role in homologous recombination-dependent DNA repair and is essential for genome stability (Xu et al., 2008 [PubMed 18923082]).[supplied by OMIM
Sequence: GKYVMVMGVVQACSPEPCLQAVKMTDLSDNPIHESMWELEVEDLHRNISpecifications
RMI2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant RMI2. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
BLAP18/C16orf75 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
116028 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
RMI2 | |
RMI2 | |
Recombinant protein corresponding to amino acids of human RMI2. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only