Learn More
ROCK2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ROCK2.
Manufacturer:  Abnova Corporation PAB12730
Catalog No. 89-116-060
Add to cart
Description
Description
The protein encoded by this gene is a serine/threonine kinase that regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element. This protein, which is an isozyme of ROCK1 is a target for the small GTPase Rho. [provided by RefSeq
Specifications
Specifications
ROCK2
Unconjugated
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
ROCK2
KIAA0619
Rabbit
100 ug
Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody recognizes ~165 KDa of human ROCK2 without a phosphorylation site ofThr396. It does not react to phosphorylated protein.
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of ROCK2.
In TBS, pH 7.2 (BSA, 10% Proclin300)
Liquid
ROCK2
A synthetic peptide corresponding to residues surrounding T396 of human ROCK2.
RUO
Primary
9475
Human, Murine, Rat