Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS6KA1 (phospho S352), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Rps6ka1.

Manufacturer:  Abnova Corporation PAB12168

Catalog No. 89-115-663


Add to cart

Description

Description

Sequence: RDSpPG
Specifications

Specifications

RPS6KA1
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Rps6ka1.
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Liquid
Rps6ka1
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S352 of mouse Rps6ka1.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
p90RSK (Phospho-Ser352) antibody detects endogenous levels of p90RSK only when phosphorylated at serine 352.
Western Blot
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Rps6ka1
Rsk1|p90rsk|rsk
Rabbit
Immunoaffinity purification
RUO
Primary
20111
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only