Learn More
RPS6KB1 (phospho T421), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RPS6KB1.
Manufacturer: Abnova Corporation PAB25842
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates several residues of the S6 ribosomal protein. The kinase activity of this protein leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding this gene and overexpression of this kinase are seen in some breast cancer cell lines. Alternate translational start sites have been described and alternate transcriptional splice variants have been observed but have not been thoroughly characterized. [provided by RefSeq
Sequence: P-R-T-P-VSpecifications
RPS6KB1 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RPS6KB1. | |
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
Liquid | |
RPS6KB1 | |
Synthetic phosphopeptide corresponding to residues surrounding T421 of human RPS6KB1. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine, Rat |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
RPS6KB1 | |
PS6K|S6K|S6K1|STK14A|p70(S6K)-alpha|p70-S6K|p70-alpha | |
Rabbit | |
Affinity chromatography | |
RUO | |
Primary | |
6198 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only