Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS6KC1, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of RPS6KC1.

Manufacturer:  Abnova Corporation PAB17438

Catalog No. 89-119-095


Add to cart

Description

Description

Specifications

Specifications

RPS6KC1
Unconjugated
Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user.
RPS6KC1
RPK118|humS6PKh1
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects endogenous levels of total RPS6KC1 protein.
ELISA, Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of RPS6KC1.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Liquid
RPS6KC1
A synthetic peptide corresponding to internal of human RPS6KC1.
RUO
Primary
26750
Human, Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only