Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-RRP12, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236547

Catalog No. NBP236547


Add to cart

Description

Description

RRP12 Polyclonal antibody specifically detects RRP12 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative).
Specifications

Specifications

RRP12
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation
NP_055994.2
FLJ20231, KIAA0690DKFZp762P1116, ribosomal RNA processing 12 homolog (S. cerevisiae), RRP12-like protein
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
RRP12
The immunogen recognized by the antibody maps to a region between residue 1247 to 1297 of human Ribosomal RNA Processing 12 Homolog using the numbering given in entry NP_055994.2.
Immunogen affinity purified
RUO
Primary
23223
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit