Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTN4 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of RTN4.

Manufacturer:  Abnova Corporation PAB13261

Catalog No. 89-116-509


Add to cart

Description

Description

This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. The product of this gene is a potent neurite outgrowth inhibitor which may also help block the regeneration of the central nervous system in higher vertebrates. Alternatively spliced transcript variants derived both from differential splicing and differential promoter usage and encoding different isoforms have been identified. [provided by RefSeq

Specifications

Specifications

RTN4
Unconjugated
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
RTN4
ASY|NI220/250|NOGO|NOGO-A|NOGOC|NSP|NSP-CL|Nbla00271|Nbla10545|Nogo-B|Nogo-C|RTN-X|RTN4-A|RTN4-B1|RTN4-B2|RTN4-C
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of RTN4.
In PBS (0.02% sodium azide)
Liquid
RTN4
A synthetic peptide corresponding to internal region 19 amino acids of human RTN4.
RUO
Primary
57142
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only