Learn More
Rabbit anti-S100A/B, Clone: D28-E, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP130154500UL
Catalog No. NBP13015450
Add to cart
Description
Description
S100A/B Monoclonal antibody specifically detects S100A/B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
S100A/B
D28-E
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:100-1:200
P23297
protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
Rabbit
IgG
500 ul
Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience
Primary
6271
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
Affinity Purified
S100A1
Peptide derived from C-terminal sequence of human S100A/B protein.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
This antibody detects S100A and S100B monomers, the alpha-beta heterodimer, and also the beta-beta homodimer