Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A6, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against full-length recombinant S100A6.

Manufacturer:  Abnova Corporation PAB12046

Catalog No. 89-115-554


Add to cart

Description

Description

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma. [provided by RefSeq

Specifications

Specifications

S100A6
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
S100A6
2A9|5B10|CABP|CACY|PRA
Rabbit
100 uL
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
Western Blot
Rabbit polyclonal antibody raised against full length recombinant S100A6.
In antiserum (0.09% sodium azide)
Liquid
S100A6
Recombinant protein corresponding to full length human S100A6.
RUO
Primary
6277
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only