Learn More
SAE1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SAE1.
Supplier:  Abnova Corporation PAB4352
Catalog No. 89-131-376
Add to Cart
Description
Description
Specifications
Specifications
SAE1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SAE1.
In PBS (0.09% sodium azide)
AOS1/FLJ3091/HSPC140/SUA1
Rabbit
Protein G purification
RUO
10055
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
SAE1
SAE1
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human SAE1.
400 μL
Primary
Human, Mouse
Liquid
Product Suggestions
Product Suggestions