Learn More
Rabbit anti-Salivary Amylase Alpha, Biotin, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP1777110.1MG
Catalog No. NBP17771101
Add to cart
Description
Description
Alpha Amylase Polyclonal antibody specifically detects Alpha Amylase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
Specifications
Alpha Amylase
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Purified
Rabbit
IgG
0.1 mg
Store at 4C. Do not freeze.
Polyclonal
Bacteria
ELISA, Immunoprecipitation, Western Blot
Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
P00692
AMY1A
a-Amylase [Bacillus amyloliquefaciens]
Delipidation and Defibrination
RUO
Primary
9780104