Learn More
SATB1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SATB1.
Manufacturer:  Abnova Corporation PAB13379
Catalog No. 89-116-627
Add to cart
Description
Description
Specifications
Specifications
SATB1
Unconjugated
Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user.
SATB1
SATB1
A synthetic peptide corresponding to C-terminus 15 amino acids of human SATB1.
RUO
Primary
6304
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of SATB1.
In PBS (0.02% sodium azide)
Liquid
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat