Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCD1 Rabbit anti-Thale cress, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of SCD1.

Manufacturer:  Abnova Corporation PAB15991

Catalog No. 89-118-001


Add to cart

Description

Description

Specifications

Specifications

SCD1
Unconjugated
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
SCD1
F27J15.17|F27J15_17|STOMATAL CYTOKINESIS-DEFECTIVE 1
Rabbit
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Thale cress
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of SCD1.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Liquid
SCD1
A synthetic peptide corresponding to C-terminus of Arabidopsis thaliana SCD1.
RUO
Primary
841327
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only