Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCN1A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Scn1a.
Supplier: Abnova Corporation PAB11759
Specifications
SCN1A | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user. | |
Scn1a | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
81574 | |
Store at 4°C. |
Immunohistochemistry (PFA fixed) | |
Lot specific | |
Rabbit polyclonal antibody raised against synthetic peptide of Scn1a. | |
In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide) | |
Scn1a | |
A synthetic peptide corresponding to amino acids 1491-1508 of rat Scn1a. | |
100 μL | |
Primary | |
Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction