Learn More
Semg1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Semg1.
Manufacturer:  Abnova Corporation PAB14672
Catalog No. 89-137-951
Add to cart
Description
Description
Specifications
Specifications
Semg1
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:5000) Immunoprecipitation (1:50-1:200) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Liquid
Native purified rat Semg1.
RUO
Primary
Human, Monkey, Murine, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Rabbit polyclonal antibody raised against native Semg1.
In serum (0.05% sodium azide)
Rabbit
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal