Learn More
SGK1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SGK1.
Manufacturer:  Abnova Corporation PAB18136
Catalog No. 89-119-709
Add to cart
Description
Description
This gene encodes a serine/threonine protein kinase that plays an important role in cellular stress response. This kinase activates certain potassium, sodium, and chloride channels, suggesting an involvement in the regulation of processes such as cell survival, neuronal excitability, and renal sodium excretion. High levels of expression of this gene may contribute to conditions such as hypertension and diabetic nephropathy. Several alternatively spliced transcript variants encoding different isoforms have been noted for this gene. [provided by RefSeq
Specifications
Specifications
SGK1
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user.
SGK1
SGK1
A synthetic peptide corresponding to human SGK1.
100 μg
Primary
This antibody is specific to SGK1.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of SGK1.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
SGK
Rabbit
Affinity purification
RUO
6446
Human, Mouse, Rat
Liquid