Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-SHP-2/PTPN11, (Human and Virus), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242920

Catalog No. NBP242920


Add to cart

Description

Description

Specifically detects SHP-2/PTPN11 in Human, Virus samples, and it is validated for ELISA, Immunocytochemistry, Immunofluorescence, Western Blotting

SHP-2/PTPN11 Polyclonal antibody specifically detects SHP-2/PTPN11 in Viral samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

SHP-2/PTPN11
Unconjugated
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Virus
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Western Blot 1:500-1:3000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:1000-1:2000
Affinity Purified
PTPN11
The immunogen is raised against SHP-2/PTPN11. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
5781
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only