Learn More
SIGLEC7 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC7.
Supplier:  Abnova Corporation PAB3650
Catalog No. 89-131-106
Add to Cart
Description
Description
Specifications
Specifications
SIGLEC7
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC7.
In PBS (0.09% sodium azide)
AIRM1/CD328/CDw328/D-siglec/QA79/SIGLEC-7/p75/p75/AIRM1
Rabbit
Protein G purification
RUO
27036
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry, Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
SIGLEC7
SIGLEC7
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SIGLEC7.
400 μL
Primary
Human
Liquid
Recommendations
Recommendations