Learn More
SIX5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SIX5.
Supplier: Abnova Corporation PAB23896
Description
The protein encoded by this gene is a homeodomain-containing transcription factor that appears to function in the regulation of organogenesis. This gene is located downstream of the dystrophia myotonica-protein kinase gene. Mutations in this gene are a cause of branchiootorenal syndrome type 2. [provided by RefSeq
Sequence: VVPTSQVVTLPQAVGPLQLLAAGPGSPVKVAAAAGPANVHLINSGVGVTALQLPSATAPGNFLLANPVSGSPIVTGVAVQQGKIILTATFPTSMLVSQVLPSpecifications
SIX5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant SIX5. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
BOR2/DMAHP | |
Rabbit | |
Antigen affinity purification | |
RUO | |
147912 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
SIX5 | |
SIX5 | |
Recombinant protein corresponding to amino acids of human SIX5. | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only